missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATM Antibody (3G11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26494-50ul
This item is not returnable.
View return policy
Description
ATM Monoclonal antibody specifically detects ATM in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| ATM | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 3G11 | |
| Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
| A synthesized peptide derived from Human ATM [UniProt Q13315] | |
| 50 μL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Protein Kinase, Tumor Suppressors | |
| 472 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction