missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlastin-2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78733PE
This item is not returnable.
View return policy
Description
Atlastin-2 Polyclonal antibody specifically detects Atlastin-2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| Atlastin-2 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| ADP-ribosylation factor-like 6 interacting protein 2, ADP-ribosylation factor-like protein 6-interacting protein 2, ADP-ribosylation-like factor 6 interacting protein 2, aip-2, ARL3IP2, ARL-6-interacting protein 2, ARL6IP2, atlastin GTPase 2, atlastin2, atlastin-2, EC 3.6.5.-, FLJ23293 | |
| The immunogen for this product maps to a region between residue 533 and 583 of human Atlastin GTPase 2 using the numbering given in entry NP_001129145.1 (GeneID 64225). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 64225 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction