missing translation for 'onlineSavingsMsg'
Learn More

Atlastin-2 Antibody, Novus Biologicals™

Product Code. 30231380 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30231380 100 μL 100µL
30232658 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30231380 Supplier Novus Biologicals Supplier No. NBP335616100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Atlastin-2 Polyclonal antibody specifically detects Atlastin-2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atlastin-2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias ADP-ribosylation factor-like 6 interacting protein 2, ADP-ribosylation factor-like protein 6-interacting protein 2, ADP-ribosylation-like factor 6 interacting protein 2, aip-2, ARL3IP2, ARL-6-interacting protein 2, ARL6IP2, atlastin GTPase 2, atlastin2, atlastin-2, EC 3.6.5.-, FLJ23293
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-470 of human Atlastin-2 (NP_001129145.1).,, Sequence:, ARDTYCKSMEQVCGGDKPYIAPSDLERKHLDLKEVAIKQFRSVKKMGGDEFCRRYQDQLEAEIEETYANFIKHNDGKNIFY
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64225
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.