Learn More
Specac™ Atlas™ Water Cooled Heated Platens and Controller
Product Code. 15280168
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
15280168 100 mm Each
1 options
Product Code. 15280168 Supplier Specac™ Supplier No. GS15515
Only null left
Add to basket
Add to basket
A quick and simple accessory for converting standard Hydraulic Presses to heatable versions. These have been designed to provide heatable pressing surfaces compatible with all Specac Presses. The Platens have a 100mm diameter pressing surface , controlled temperature capability up to 300°C and a 15 ton load bearing capacity.
Specifications
Diameter (Metric) 100 mm