Learn More
Specac™ Atlas™ Manual 15T Hydraulic Press
Product Code. 12960880
Encompass_Preferred
Change view
Click to view available options
Type:
Atlas Manual 15T Hydraulic Press
Unit Size:
Each
Product Code. Type Unit Size
12960880 Atlas Manual 15T Hydraulic Press Each
1 options
Product Code. 12960880 Supplier Specac™ Supplier No. GS15011
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manual Hydraulic 15 Ton Press. Designed to handle a wide variety of pressing applications. Specifically suited to preparation of Kbr discs using range of Evacuable Pellet Dies. Can also be used with Heated Platens for applications such as preparation of thin polymer films.
Specifications
For Use With (Equipment) Heated Platens for Applications Such as Preparation of Thin Polymer Films.
Type Atlas Manual 15T Hydraulic Press