Learn More
Specac™ Atlas™ Hydraulic Press Manual 25T
Product Code. 15260328
Encompass_Preferred
Change view
Click to view available options
Type:
Atlas Hydraulic Press Manual 25T
Unit Size:
Each
Product Code. Type unitSize
15260328 Atlas Hydraulic Press Manual 25T Each
1 options
Product Code. 15260328 Supplier Specac™ Supplier No. GS25011
Only null left
Add to basket
Add to basket
This is a Manual Hydraulic 25 Ton Press designed to handle a wide variety of pressing applications. Although specifically suited to the preparation of KBr discs using the Specac Evacuable Pellet Die Assembly this Press can turn any powder into a pellet.
Specifications
Type Atlas Hydraulic Press Manual 25T