missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Atlas™ Hydraulic Press Automatic 8T Korea 220V
Click to view available options
Type:
Atlas Hydraulic Press Automatic 8T Korea 220V
Description
This Atlas Power 8T Press is a power assisted hydraulic press that operates at up to 8 Tons. It has been designed to handle a wide variety of pressing applications including XRF and IR sample preparation. This press has a microprocessor controlled pressure application and release, Touch Screen operation and can accommodate samples up to 20mm in diameter. They can also be used with Specac Heated Platens GS15515 and Specac Evacuable Pellet Dies. Supplied with Korean Plug.
Specifications
Specifications
| Type | Atlas Hydraulic Press Automatic 8T Korea 220V |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction