missing translation for 'onlineSavingsMsg'
Få mere at vide

Specac™ Atlas™ Hydraulic Press Automatic 25T EU 230V

Artikelnummer. 15220348
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Type:
Atlas Hydraulic Press Automatic 25T EU 230V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Type
15220348 Atlas Hydraulic Press Automatic 25T EU 230V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15220348 Leverandør Specac™ Leverandørnr. GS25420
 Restricted Item
Estimated Shipment: 16-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
This Atlas Power 25T Press is a power assisted hydraulic press that operates at up to 25 Tons. It has been designed to handle a wide variety of pressing applications including XRF and IR sample preparation. This press has a microprocessor controlled pressure application and release, Touch Screen operation and can accommodate samples up to 200mm in diameter. They can also be used with Specac Heated Platens GS15515 and Specac Evacuable Pellet Dies. Supplied with UK/EU Plug.

Tekniske data

Type Atlas Hydraulic Press Automatic 25T EU 230V
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.