Learn More
Specac™ Atlas™ High Temperature Constant Thickness Film-maker
Product Code. 15290178
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
29 mm
Product Code. Diameter (Metric)
15290178 29 mm
1 options
Product Code. 15290178 Supplier Specac™ Supplier No. GS15800
Add to basket
Add to basket
Atlas High Temperature Film Maker Kit. This kit is a film making system to produce high quality, reproducible polymer films using temperatures up to 400°C. This product has a set of heated platens already incorporated into the film making part of the accessory and so Heated Platens GS15515 are not required. Can produce 6 different thickness of film, all of 29mm diameter.
Specifications
Description Can produce 6 different thickness of film, all of 29 mm diameter
Diameter (Metric) 29 mm