missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Atlas™ Evacuable Pellet Die 10mm
Click to view available options
Diameter (Metric):
10 mm
Unit Size:
Each
Description
10mm Evacuable Pellet Die used for compacting powdered samples into discs or briquettes with use of a pressing system. Well suited for KBr pellet production for FTIR analysis and XRF pellets. This product comprises of a base, body, plunger, pair of pressing surfaces, extractor ring + O-Ring Kit.
Specifications
Specifications
| Diameter (Metric) | 10 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction