missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566790
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: RVAAAASRGAD DAMESSKPGP VQVVLVQKDQ HSFEL Highest antigen sequence identity to the following orthologs - mouse 81%, rat 58%.
ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.Specifications
ATL3 | |
Polyclonal | |
Unconjugated | |
Atl3 | |
4633402C03Rik; 5730596K20Rik; AI465397; Atl3; atlastin 3; atlastin GTPase 3; Atlastin-3; AW228836; HSN1F; I79_021367; RGD1309871 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25923 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DD88 | |
Atl3 | |
Recombinant Human ATL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only