Learn More
ATL3 Mouse, anti-Human, Clone: 2E7C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68516-1-IG-150UL
Description
The ATL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATL3. This antibody recognizes human, mouse, rat, rabbit antigen. The ATL3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ATL3 | |
| 2E7C4 | |
| ATL3, Atlastin 3, atlastin GTPase 3, DKFZp564J0863 | |
| 150 μL | |
| 25923 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG1 |