missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556058
This item is not returnable.
View return policy
Description
Immunogen sequence: EFREIGTVID QIAETLWEQR SPRKVFSKLF EVTRRRMVHR ALSSAQRQRL SSNNNKKK Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 40%.
ATL2 gene ontology annotations related to this gene include GTPase activity; GTP binding.
Specifications
| ATL2 | |
| Polyclonal | |
| Unconjugated | |
| ATL2 | |
| 2010110I21Rik; AA407293; ADAMTSL-2; ADP-ribosylation factor-like 6 interacting protein 2; ADP-ribosylation factor-like protein 6-interacting protein 2; ADP-ribosylation-like factor 6 interacting protein 2; aip-2; ARL3IP2; ARL-6 interacting protein-2; ARL-6-interacting protein 2; ARL6IP2; Atl2; atlastin GTPase 2; atlastin GTPase 2; atlastin-2; atlastin2; atlastin-2; AV334690; fc06e01; GPHYSD1; wu:fc06e01; wu:fc06e01 protein; wu:fv60d03 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64225 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NHH9 | |
| ATL2 | |
| Recombinant protein corresponding to Human ATL2. Recombinant protein control fragment (Product #RP-90939). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction