missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576706
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| ATL1 | |
| Polyclonal | |
| Unconjugated | |
| ATL1 | |
| 4930435M24Rik; Adfsp; AD-FSP; ATL1; ATL1 alpha; ATL1 beta; ATL1 gamma; ATL1-delta; atlastin GTPase 1; atlastin1; atlastin-1; Bcl11B; BCL-11B; BCL11B/TRDC fusion; brain-specific GTP-binding protein; Ctip 2; CTIP2; CTIP-2; Fsp1; GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate-binding protein 3; hGBP3; hRit1; hRIT1 alpha; HSN1D; Rit 1; Rit1; spastic paraplegia 3 protein A; Spastic paraplegia 3A homolog; SPG3; SPG3A; ZNF856B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51062, 73991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BH66, Q8WXF7 | |
| ATL1 | |
| Recombinant full length Human ATL1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction