missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATL1 Monoclonal Antibody (1F6B12)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531641

Product Code. 16345074

  • £431.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-31641 has been tested in indirect ELISA.

The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATL1
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q8WXF7
ATL1
Purified recombinant fragment of human ATL1 (AA: 1-100) expressed in E. coli.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Western Blot
1F6B12
Unconjugated
ATL1
4930435M24Rik; Adfsp; AD-FSP; ATL1; ATL1 alpha; ATL1 beta; ATL1 gamma; ATL1-delta; atlastin GTPase 1; atlastin1; atlastin-1; Bcl11B; BCL-11B; BCL11B/TRDC fusion; brain-specific GTP-binding protein; Ctip 2; CTIP2; CTIP-2; Fsp1; GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate-binding protein 3; hGBP3; hRit1; hRIT1 alpha; HSN1D; Rit 1; Rit1; spastic paraplegia 3 protein A; Spastic paraplegia 3A homolog; SPG3; SPG3A; ZNF856B
Mouse
Protein G
RUO
51062
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.