missing translation for 'onlineSavingsMsg'
Learn More

Atipamezole Hydrochloride, TRC

Product Code. 30011417
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011417 100 mg 100mg
30010911 10 mg 10mg
30009092 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011417 Supplier Toronto Research Chemicals Supplier No. TRCA790100100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104075-48-1
Molecular Formula C14 H16 N2 . Cl H
Molecular Weight (g/mol) 248.75
Synonym Atipamezole Hydrochloride, 1H-Imidazole, 4-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-, monohydrochloride (9CI), Antisedan, Atipamezole hydrochloride, 1H-Imidazole, 5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-, hydrochloride (1:1), 4-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole monohydrochloride
IUPAC Name 4-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole;hydrochloride
SMILES Cl.CCC1(Cc2ccccc2C1)c3c[nH]cn3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H16N2.ClH/c1-2-14(13-9-15-10-16-13)7-11-5-3-4-6-12(11)8-14;/h3-6,9-10H,2,7-8H2,1H3,(H,15,16);1H
Formula Weight 248.108
Percent Purity >95%
Chemical Name or Material Atipamezole Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.