missing translation for 'onlineSavingsMsg'
Learn More

Atipamezole hydrochloride, MedChemExpress

Product Code. 16475128
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475128 10 mg 10mg
16421367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475128 Supplier Medchem Express Supplier No. HY12380
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Atipamezole (MPV-1248) hydrochloride is a potent α2-adrenoceptor antagonist with a Ki of 1.6 nM.

Atipamezole (MPV-1248) hydrochloride is a potent α2-adrenoceptor antagonist with a Ki of 1.6 nM.

Specifications

Chemical Name or Material Atipamezole hydrochloride
CAS 104075-48-1
Color White
Purity Grade Notes Research
Molecular Formula C14H17ClN2
Quantity 10 mg
Synonym MPV-1248 hydrochloride
Solubility Information H2O : 75 mg/mL (301.51 mM; Need ultrasonic and warming) ∣DMSO : ≥ 47 mg/mL (188.94 mM)
SMILES CCC1(C2=CN=CN2)CC3=C(C=CC=C3)C1.[H]Cl
Molecular Weight (g/mol) 248.75
Formula Weight 248.75
Percent Purity 97.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.