Learn More
Invitrogen™ ATIC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117021
Description
Positive Samples: MCF7, SW480, NCI-H460, HepG2, SW620, HT-29, K-562 Immunogen sequence: MAPGQLALFS VSDKTGLVEF ARNLTALGLN LVASGGTAKA LRDAGLAVRD VSELTGFPEM LGGRVKTLHP AVHAGILARN IPEDNADMAR LDFNLIRVVA CNLYPFVKTV ASPGVTVEEA VEQIDIGGVT LLRAAAKNHA RVTVVCEPED YVVVSTEMQS SESKDTSLET RRQLALKAFT HTAQYDEAIS DYFRKQYSKG VSQMPLRYGM NPHQTPAQLY TLQPKLPITV LNGAPGFINL CDALNAWQLV KELKEALGIP AAASFKHVSP AGAAVGIPLS EDEAKVCMVY DLYKTLTPIS.
ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.
Specifications
| ATIC | |
| Polyclonal | |
| Unconjugated | |
| ATIC | |
| 153761_at; 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; atic; atic.L; AW212393; BEST:GH11240; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; CG11089; CG11089-PA; CG11089-PB; CT30991; Dmel\CG11089; Dmel_CG11089; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; RE29555p; uncharacterized protein; XELAEV_18044849mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 471 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31939 | |
| ATIC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ATIC (NP_0040352). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.