missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ATI 2341

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
ATI 2341 is a CXCR4 allosteric agonist (EC50 = 194 nM). Induces chemotaxis in CXCR4-expressing cells in vitro. Mobilizes polymorphonuclear neutrophils (PMNs) and hematopoietic stem and progenitor cells (HSPCs) from bone marrow niche in vivo. Exhibits biased signaling towards Gi proteins over G13 and β-arrestin.
Specifications
Specifications
| Purity | 0.95 |
| CAS | 1337878-62-2 |
| Content And Storage | Store at -20°C |
| Description | Chemokine Receptor Agonists |
| Molecular Weight (g/mol) | Observed M.W.: 2256.82 |
| Quantity | 1 mg |
| Structural Form | C104H178N26O25S2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction