missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atherin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04030JF549
This item is not returnable.
View return policy
Description
Atherin Polyclonal antibody specifically detects Atherin in Human samples. It is validated for Western Blot
Specifications
| Atherin | |
| Polyclonal | |
| Western Blot | |
| SAMD1 sterile alpha motif domain containing 1 | |
| The immunogen recognized by this antibody maps to this antibody Motif Domain Containing 1 using the numbering given in entry NP_612361.1 (GeneID 90378). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 90378 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction