missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATGL Recombinant Rabbit Monoclonal Antibody (10Y4M10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538135
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, A-431, U-87MG, Mouse brain, Mouse kidney, Mouse heart muscle, Rat brain, Rat kidney, Rat heart muscle Immunogen sequence: LPARLNEALL EACVEPTDLL TTLSNMLPVR LATAMMVPYT LPLESALSFT IRLLEWLPDV PEDIRWMKEQ TGSICQYLVM RAKRKLGRHL PSRLP.
Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
Specifications
| ATGL | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C548, Q8BJ56, Q96AD5 | |
| PNPLA2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-394 of human ATGL / PNPLA2 (NP_0651091). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 10Y4M10 | |
| Unconjugated | |
| PNPLA2 | |
| 0610039C21Rik; 1110001C14Rik; adipose triglyceride lipase; ATGL; ATGL DESNUTRIN; calcium-independent phospholipase A2; Desnutrin; DKFZp667M109; FP17548; IPLA2 zeta; iPLA2zeta; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; patatin-like phospholipase domain-containing protein 2; PEDF R; PEDF-R; pigment epithelium-derived factor; plpl; plpl2; Pnpla2; RGD1309044; transport-secretion protein; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS 2.2; TTS2; TTS2.2; TTS-2.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361676, 57104, 66853 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction