Learn More
ATGL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55190-1-AP-150UL
 View more versions of this product
Product Code. 16876675
  • £349.00 / 150µL
Description
This antibody is specific to ATGL.
Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
Specifications
Specifications
ATGL
Polyclonal
Unconjugated
PNPLA2
1110001C14Rik, Adipose Triglyceride Lipase, ATGL, DESNUTRIN, FP17548, IPLA2 zeta, PEDF R, PNPLA2, Transport secretion protein 2, TTS 2.2, TTS2, TTS2.2
Rabbit
Antigen Affinity Chromatography
RUO
361676, 57104, 66853
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P0C548, Q8BJ56, Q96AD5
Pnpla2
Peptide
150 μL
Primary
Human, Mouse, Rat