missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9B Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77169AF488
This item is not returnable.
View return policy
Description
ATG9B Polyclonal antibody specifically detects ATG9B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ATG9B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ATG9B. The immunogen is located within amino acids 850 - 900 of ATG9B. Amino Acid Squence: CPSQPPSPDEEKPSWSSD | |
| 0.1 mL | |
| Autophagy, Golgi Apparatus Markers | |
| 285973 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction