missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG9A Polyclonal Antibody
GREENER_CHOICE

Product Code. 12819192
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12819192 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12819192 Supplier Invitrogen™ Supplier No. PA521043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse heart tissue lysate. PA5-21043 can be used with blocking peptide PEP-1157.

Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Apg9 plays a direct role in the formation of the cytoplasm to vacuole targeting and autophagic vesicles, possibly serving as a marker for a specialized compartment essential for these vesicle-mediated alternative targeting pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG9A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ATG9A
Gene Accession No. Q5FWU3, Q68FE2, Q7Z3C6
Gene Alias Apg9; APG9 autophagy 9-like 1; Apg911; APG9A; APG9L1; APG9-like 1; ATG 9A; Atg9; ATG9 autophagy related 9 homolog A; ATG9 autophagy related 9 homolog A (S. cerevisiae); Atg9a; Atg9l1; AU019532; autophagy 9-like 1 protein; autophagy protein 9; autophagy related 9A; autophagy-related 9A; autophagy-related 9-like 1; autophagy-related protein 9; autophagy-related protein 9A; FLJ22169; mATG9; MGD3208; RGD1310450; zgc:158700
Gene Symbols ATG9A
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human ATG9A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245860, 363254, 79065
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.