missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9A Monoclonal Antibody (14F2 8B1), Alexa Fluor™ 647, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA1149A647
This item is not returnable.
View return policy
Description
MA1-149-A647 was produced in Armenian hamster and detects ATG9 in human, rat and mouse samples. In Immunofluorescence applications, MA1-149A-647 shows accumulation and redestribution of ATG9 in response to starvation-induced autophagosome assembly.
ATG9A Monoclonal antibody specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry
Specifications
| ATG9A | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1.0mg/mL BSA and 0.05% sodium azide | |
| Q5FWU3, Q68FE2, Q7Z3C6 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 245860, 363254, 79065 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG |
| Immunocytochemistry | |
| 14F2 8B1 | |
| Alexa Fluor 647 | |
| ATG9A | |
| Apg9; APG9 autophagy 9-like 1; Apg911; APG9A; APG9L1; APG9-like 1; ATG 9A; Atg9; ATG9 autophagy related 9 homolog A; ATG9 autophagy related 9 homolog A (S. cerevisiae); Atg9a; Atg9l1; AU019532; autophagy 9-like 1 protein; autophagy protein 9; autophagy related 9A; autophagy-related 9A; autophagy-related 9-like 1; autophagy-related protein 9; autophagy-related protein 9A; FLJ22169; mATG9; MGD3208; RGD1310450; zgc:158700 | |
| peptide sequence HPEPVPEEGSEDELPPQVHK of human ATG9A C-terminus | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion