missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9A Monoclonal Antibody (14F2 8B1), Alexa Fluor™ 555, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA1149A555
This item is not returnable.
View return policy
Description
MA1-149A555 was produced in Armenian hamster and detects ATG9 in human, rat and mouse samples. In Immunofluorescence applications, MA1-149A555 shows accumulation and redestribution of ATG9 in response to starvation-induced autophagosome assembly.
Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Apg9 plays a direct role in the formation of the cytoplasm to vacuole targeting and autophagic vesicles, possibly serving as a marker for a specialized compartment essential for these vesicle-mediated alternative targeting pathways.
Specifications
| ATG9A | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1.0mg/mL BSA and 0.05% sodium azide | |
| Q5FWU3, Q68FE2, Q7Z3C6 | |
| ATG9A | |
| peptide sequence HPEPVPEEGSEDELPPQVHK of human ATG9A C-terminus | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Western Blot | |
| 14F2 8B1 | |
| Alexa Fluor 555 | |
| ATG9A | |
| Apg9; APG9 autophagy 9-like 1; Apg911; APG9A; APG9L1; APG9-like 1; ATG 9A; Atg9; ATG9 autophagy related 9 homolog A; ATG9 autophagy related 9 homolog A (S. cerevisiae); Atg9a; Atg9l1; AU019532; autophagy 9-like 1 protein; autophagy protein 9; autophagy related 9A; autophagy-related 9A; autophagy-related 9-like 1; autophagy-related protein 9; autophagy-related protein 9A; FLJ22169; mATG9; MGD3208; RGD1310450; zgc:158700 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 245860, 363254, 79065 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction