missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATG9A Antibody (SR3028), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | ATG9A |
|---|---|
| Clone | SR3028 |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041549
|
Novus Biologicals™
NBP3-27739-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041522
|
Novus Biologicals™
NBP3-27739-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG9A Monoclonal antibody specifically detects ATG9A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| ATG9A | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
| A synthesized peptide derived from human ATG9A (Uniprot #: Q7Z3C6) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3028 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 79065 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title