missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9A Antibody (Atg9 14F2 8B1), FITC, Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-50109F
This item is not returnable.
View return policy
Description
ATG9A Monoclonal specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
| ATG9A | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ATG9A | |
| Conserved C-terminal synthetic peptide of Human ATG9A sequence | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 79065 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Atg9 14F2 8B1 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
| Hamster | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction