missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9A Antibody (Atg9 14F2 8B1) - BSA Free, Novus Biologicals™
Hamster Monoclonal Antibody
£154.00 - £314.00
Specifications
| Antigen | ATG9A |
|---|---|
| Clone | Atg9 14F2 8B1 |
| Concentration | 0.7 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Knockdown Validated |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636406
|
Novus Biologicals
NBP2-50109-0.025ml |
0.025 mL |
£154.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643977
|
Novus Biologicals
NBP2-50109 |
0.1 mL |
£314.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG9A Monoclonal antibody specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDownSpecifications
| ATG9A | |
| 0.7 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Hamster | |
| Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| PBS | |
| 79065 | |
| IgG | |
| Protein G purified |
| Atg9 14F2 8B1 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
| Conserved C-terminal synthetic peptide of Human ATG9A sequence | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title