missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG7 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
£429.00
Specifications
| Antigen | ATG7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Form | Purified |
Description
ATG7 Polyclonal antibody specifically detects ATG7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ATG7 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human APG7. The immunogen is located within the first 50 amino acids of APG7. Amino Acid Squence: QKKLNEYRLDEAPKDIK | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| PerCP | |
| Rabbit | |
| Autophagy | |
| PBS | |
| 10533 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title