missing translation for 'onlineSavingsMsg'
Learn More

ATG7 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18422366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422366 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422366 Supplier Novus Biologicals Supplier No. NBP176650JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG7 Polyclonal antibody specifically detects ATG7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG7
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human APG7. The immunogen is located within the first 50 amino acids of APG7. Amino Acid Squence: QKKLNEYRLDEAPKDIK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 10533
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.