missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 Mouse anti-Human, Clone: ATG5/2492, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | ATG5 |
|---|---|
| Clone | ATG5/2492 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA 1-5 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Description
ATG5 Monoclonal specifically detects ATG5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
| ATG5 | |
| 0.2 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 9474 | |
| IgG1 κ | |
| Protein A or G purified |
| ATG5/2492 | |
| Western Blot 1-2 ug/ml, ELISA 1-5 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
| Recombinant fragment of human ATG5 protein (around aa 1-119) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title