missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 Mouse anti-Human, Clone: ATG5/2492, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07879-100ug
This item is not returnable.
View return policy
Description
ATG5 Monoclonal specifically detects ATG5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications
| ATG5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, ELISA 1-5 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25 | |
| Recombinant fragment of human ATG5 protein (around aa 1-119) (exact sequence is proprietary) | |
| 100 μg | |
| Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway | |
| 9474 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| ATG5/2492 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu