missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4C Monoclonal Antibody (287CT12.2.2)

ATG4C Monoclonal Antibody (287CT12.2.2)
Brand: Thermo Scientific MA537526
This item is not returnable.
View return policy
Description
ATG4C Monoclonal Antibody for Western Blot
Specifications
| ATG4C | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q811C2 | |
| ATG4C | |
| ATG4C recombinant protein. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 287CT12.2.2 | |
| Unconjugated | |
| ATG4C | |
| APG4 (ATG4) autophagy-related homolog C; APG4 autophagy 4 homolog C; APG4C; APG4-C; ATG4 autophagy related 4 homolog C; ATG4C; Atg4cl; AUTL1; AUTL3; AUT-like 1, cysteine endopeptidase; AUT-like 3 cysteine endopeptidase; autophagin 3; autophagin-3; autophagy related 4C cysteine peptidase; autophagy related 4C, cysteine peptidase; autophagy-related 4C; autophagy-related cysteine endopeptidase 3; Autophagy-related protein 4 homolog C; cysteine peptidase; cysteine protease ATG4C; cysteine protease involved in autophagy; FLJ14867 | |
| Mouse | |
| Protein G | |
| RUO | |
| 242557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction