missing translation for 'onlineSavingsMsg'
Learn More

ATG4C Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18426366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426366 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426366 Supplier Novus Biologicals Supplier No. NBP281954JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG4C Polyclonal antibody specifically detects ATG4C in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4C
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias APG4 autophagy 4 homolog C, APG4 autophagy 4 homolog C (S. cerevisiae), APG4C, ATG4 autophagy related 4 homolog C (S. cerevisiae), AUT (S. cerevisiae)-like 1, cysteine endopeptidase; AUT-like 1, cysteineendopeptidase (S. cerevisiae), AUTL1, AUTL3APG4-C, AUT-like 1, cysteine endopeptidase, AUT-like 3 cysteine endopeptidase, Autophagin-3, Autophagy-related cysteine endopeptidase 3, Autophagy-related protein 4 homolog C, cysteine protease ATG4C, EC 3.4.22, EC 3.4.22.-, FLJ14867
Host Species Rabbit
Immunogen ATG4C antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ATG4C. The immunogen is located within the last 50 amino acids of ATG4C. Amino Acid Squence: NGHSRDYDFTSTTTNEED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84938
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.