missing translation for 'onlineSavingsMsg'
Learn More

ATG4B Rabbit anti-Human, Mouse, Rat, Clone: 9H6M6, Novus Biologicals™

Product Code. 18338796 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338796 100 μg 100µL
18381004 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18338796 Supplier Novus Biologicals Supplier No. NBP316538100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATG4B Monoclonal antibody specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG4B
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 9H6M6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ATG4B (Q9Y4P1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 23192
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.