missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG4A Recombinant Rabbit Monoclonal Antibody (23GB3235), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ATG4A |
|---|---|
| Clone | 23GB3235 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088984
![]() |
Invitrogen™
MA552602 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089633
|
Invitrogen™
MA552603 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG4A Recombinant Monoclonal antibody specifically detects ATG4A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ATG4A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8C9S8, Q8WYN0 | |
| 115201, 666468, 678769 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB3235 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI627006; APG4 (ATG4) autophagy-related homolog A; APG4 autophagy 4 homolog A; APG4A; ATG4 autophagy related 4 homolog A; Atg4a; Atg4al; Atg4al1; AUTL2; AUT-like 2 cysteine endopeptidase; AUT-like 2, cysteine endopeptidase; autophagin 2; autophagin-2; autophagy 4 homolog A; autophagy related 4 homolog A; autophagy related 4A cysteine peptidase; autophagy related 4A, cysteine peptidase; autophagy related 4A, cysteine peptidase-like 1; autophagy-related 4A; autophagy-related cysteine endopeptidase 2; Autophagy-related protein 4 homolog A; AV169859; cysteine protease APG4A; cysteine protease ATG4A; hAPG4A; MGC107179 | |
| A synthetic peptide of human Atg4A (1-84AA). | |
| Primary | |
| -20°C | |
| ATG4A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title