missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27467-1-AP-150UL
This item is not returnable.
View return policy
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified.Specifications
| ATG4A | |
| Polyclonal | |
| Unconjugated | |
| ATG4A | |
| APG4A, ATG4A, AUTL2, Autophagin 2, Cysteine protease ATG4A, hAPG4A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115201, 666468 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8C9S8, Q8WYN0 | |
| Atg4a | |
| ATG4A Fusion Protein Ag26497 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction