missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG3 Recombinant Rabbit Monoclonal Antibody (23GB4330), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552688
This item is not returnable.
View return policy
Description
ATG3 (APG3) is a widely expressed conjugating enzyme for APG8 lipidation, an essential step for the initiation of autophagy. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. Autophagy is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG3 functions as an E2-like enzyme during the initial stages of autophagosome formation by catalyzing the formation of the Atg8-phosphatidylethanolamine (Atg8-PE) conjugate, which is critical for autophagy.
Specifications
| ATG3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATG3 | |
| 2610016C12Rik; APG3; Apg3l; APG3-like; Apg3p; atg3; ATG3 autophagy related 3 homolog; atg3.S; Atg3l; autophagy related 3; autophagy related 3 S homeolog; autophagy-related 3; autophagy-related protein 3; DKFZp564M1178; EGK_11288; FLJ22125; hApg3; MGC15201; PC3-96; Pig1; PIG-1; preconditioning-inducible gene 1 protein; Protein PC3-96; Ubiquitin-like-conjugating enzyme ATG3; XELAEV_18014721mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171415, 64422, 67841 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4330 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AZ50, Q9CPX6, Q9NT62 | |
| ATG3 | |
| A synthesized peptide derived from human Apg3 (Atg3) (1-30AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction