missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG2B Polyclonal Antibody

Description
Keep as concentrated solution. Predicted reactivity: Mouse (82%), Pig (91%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. Another member of the autophagy family of proteins is ATG2B, one of two homologs of ATG2 that is essential for autophagosome formation and important for regulation of size and distribution of lipid droplets. Relatively high rates of ATG2B mutations were observed in gastric and colorectal carcinomas, suggesting that deregulating the autophagy process may contribute to cancer development.
Specifications
Specifications
| Antigen | ATG2B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.97 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | ATG2B |
| Gene Accession No. | Q96BY7 |
| Gene Alias | 2410024A21Rik; AI047755; AI503411; ATG2 autophagy related 2 homolog B; Atg2b; autophagy related 2B; autophagy-related protein 2 homolog B; AW558123; C030004M05Rik; C14orf103; C630028L02Rik; mKIAA4067; RGD1304878 |
| Gene Symbols | ATG2B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?