missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570081
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 80%.
Involved in autophagosome assembly, regulating the size of nascent autophagosomes (PubMed:28561066). Also regulates lipid droplets morphology and distribution within the cell (PubMed:22219374, PubMed:28561066). [UniProt].
Specifications
| ATG2A | |
| Polyclonal | |
| Unconjugated | |
| ATG2A | |
| 1810013C15Rik; A830054M12; ATG2 autophagy related 2 homolog A; Atg2a; autophagy related 2A; autophagy-related 2-like protein A; autophagy-related protein 2 homolog A; BC023754; Kiaa0404; mKIAA0404 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23130 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2TAZ0 | |
| ATG2A | |
| synthetic peptide directed towards the N terminal region of human ATG2A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction