missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG2A Antibody (4E6D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61690-0.025ml
This item is not returnable.
View return policy
Description
ATG2A Monoclonal specifically detects ATG2A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ATG2A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| ATG2 autophagy related 2 homolog A (S. cerevisiae), autophagy-related protein 2 homolog A, KIAA0404MGC117153 | |
| Mouse | |
| 212.8 kDa | |
| 0.025 mL | |
| 23130 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4E6D4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ATG2A | |
| Purified recombinant fragment of human ATG2A (AA: 325-429) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction