missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG16L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561000
This item is not returnable.
View return policy
Description
Immunogen sequence: LEKEACEKWK RPFRSASATS LTLSHCVDVV KGLLDFKKRR GHSIGGAPEQ RYQIIPVCVA ARLPTRAQDV LDAHLSEVNA VRFGPNS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
May play a role in autophagy. [UniProt].
Specifications
| ATG16L2 | |
| Polyclonal | |
| Unconjugated | |
| ATG16L2 | |
| 2410118P20Rik; APG16-like 2; ATG16 autophagy related 16-like 2; ATG16B; Atg16l2; Atg16L2 alpha; Atg16L2 beta; autophagy related 16 like 2; autophagy related 16-like 2; autophagy related 16-like 2 (S. cerevisiae); autophagy-related 16-like 2 alpha; autophagy-related 16-like 2 beta; autophagy-related protein 16-2; mFLJ00012 protein; RGD1311400; WD repeat-containing protein 80; WDR80 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NAA4 | |
| ATG16L2 | |
| Recombinant protein corresponding to Human ATG16L2. Recombinant protein control fragment (Product #RP-97441). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction