missing translation for 'onlineSavingsMsg'
Learn More

ATG16L1 Rabbit anti-Human, Mouse, Clone: 5N2N9, Novus Biologicals™

Product Code. 18352312
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352312 100 μg 100µL
18311445 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18352312 Supplier Bio-Techne Supplier No. NBP316236100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATG16L1 Monoclonal antibody specifically detects ATG16L1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen ATG16L1
Applications Western Blot
Classification Monoclonal
Clone 5N2N9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 6-105 of human ATG16L1 (Q676U5).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 55054
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.