missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L1 Mouse anti-Human, Clone:8F8C2, Abnova™
Mouse monoclonal antibody raised against recombinant human ATG16L1.
Brand: Abnova MAB17120.100ug
This item is not returnable.
View return policy
Description
Autophagy is the major intracellular degradation system delivering cytoplasmic components to lysosomes, and it accounts for degradation of most long-lived proteins and some organelles. Cytoplasmic constituents, including organelles, are sequestered into double-membraned autophagosomes, which subsequently fuse with lysosomes. ATG16L1 is a component of a large protein complex essential for autophagy (Mizushima et al., 2003 [PubMed 12665549]).[supplied by OMIM]Specifications
| ATG16L1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| ATG16L1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 8F8C2 | |
| ATG16 autophagy related 16-like 1 (S. cerevisiae) | |
| In PBS (0.05% sodium azide) | |
| APG16L, ATG16L, FLJ00045, FLJ10035, FLJ10828, FLJ22677, IBD10, WDR30 | |
| Recombinant protein corresponding to amino acid 11-257 of human ATG16L1 from E. coli. | |
| 55054 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction