missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£163.00 - £442.00
Specifications
| Antigen | ATG14 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18728873
|
Novus Biologicals
NBP2-36445-0.1mg |
0.1 mg |
£442.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647405
|
Novus Biologicals
NBP2-36445-0.025mg |
0.025 mg |
£163.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG14 Polyclonal specifically detects ATG14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ATG14 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 22863 | |
| ATG14 antibody was raised against an 18 amino acid synthetic peptide near the center of human ATG14. | |
| Primary | |
| At least three isoforms of ATG14 are known to exist; this antibody will detect all three isoforms |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831 | |
| ATG14 | |
| IgG | |
| Affinity Purified | |
| 54 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title