missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATG14 Antibody (1F7C10), Novus Biologicals™

Mouse Monoclonal Antibody

£217.00 - £389.00

Specifications

Antigen ATG14
Clone 1F7C10
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30029187
View Documents
Novus Biologicals™
NBP3-27119-0.1ml
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30039759
View Documents
Novus Biologicals™
NBP3-27119-0.025ml
0.025 mL
£217.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATG14 Monoclonal antibody specifically detects ATG14 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

ATG14
1 mg/mL
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Autophagy, Cell Biology, Endocrinology, Signal Transduction
PBS
22863
IgG1
Protein G purified
1F7C10
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Monoclonal
Purified
RUO
Human
ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831
Purified recombinant fragment of human ATG14 (AA: 43-303) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.