Learn More
Invitrogen™ ATG13 Recombinant Rabbit Monoclonal Antibody (K01_1K67)
Description
Immunogen sequence: QTVRVGTVGT PVGTITLSCA YRINLAFMST RQFERTPPIM GIIIDHFVDR PYPSSSPMHP CNYRTAGEDT GVIYPSVEDS QEVCTTSFST SPPSQLSSSR LSYQPAALGV GSADLAYPVV FAAGLNATHP HQLMVPGKEG GVPLAPNQPV HGTQADQERL ATCTPSDRTH CAATPSSSED TETVSNSSEG RASPHDVLET IFVRKVGAFV NKPINQVTLT SLDIPFAMFA PKNLELEDTD PMVNPPDSPE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Pig - 96%, Cynomolgus monkey - 98%.
Specifications
Specifications
| Antigen | ATG13 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1K67 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | ATG13 |
| Gene Accession No. | O75143, Q91YI1 |
| Gene Alias | 1110053A20Rik; Atg13; ATG13 autophagy related 13 homolog; autophagy related 13; autophagy-related protein 13; D2Ertd391e; FLJ20698; Harbi1; Harbi1l; harbinger transposase derived 1; KIAA0652; OTTHUMP00000233321; OTTHUMP00000233322; OTTHUMP00000233323; OTTHUMP00000233324; OTTHUMP00000233325; OTTHUMP00000233326; PARATARG8; RGD1310685 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.