missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG13 Recombinant Rabbit Monoclonal Antibody (24GB6785)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556979
This item is not returnable.
View return policy
Description
ATG13 Recombinant Monoclonal Antibody for Western Blot
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein ATG1. ATG13 forms a complex with ULK1 and ULK2, the mammalian homologs of ATG1, and with FIP200. This complex is a target of TOR phosphorylation under normal conditions; inhibition of TOR by rapamycin or leucine deprivation leads to dephosphorylation of ATG13, ULK1 and ULK2, which then leads to autophagy. Knockdown of ATG13 inhibits autophagosome formation.
Specifications
| ATG13 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATG13 | |
| 1110053A20Rik; Atg13; ATG13 autophagy related 13 homolog; autophagy related 13; autophagy-related protein 13; D2Ertd391e; FLJ20698; Harbi1; Harbi1l; harbinger transposase derived 1; KIAA0652; OTTHUMP00000233321; OTTHUMP00000233322; OTTHUMP00000233323; OTTHUMP00000233324; OTTHUMP00000233325; OTTHUMP00000233326; PARATARG8; RGD1310685 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362164, 51897, 9776 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6785 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75143, Q91YI1 | |
| ATG13 | |
| Recombinant protein of human KIaa 0652 (aa 169-418). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction