missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATG12 Monoclonal antibody specifically detects ATG12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ATG12 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 9R8W2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 61-140 of human ATG12 (O94817). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?